Affinity chromatography columns, HiTrap™ Benzamidine FF
HiTrap™ Benzamidine FF (High Sub), PP, 90 µm, 7×25 mm, 1 ml, <4 ml/min
Catalog # 17-5143-01
Supplier: Cytiva
Jump to:
Product Details & Documents
HiTrap Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
- Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates
- High binding capacity
- It can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC system
HiTrap Benzamidine FF (high sub) are prepacked 1 ml and 5 ml columns for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.
The medium, Benzamidine Sepharose 4 Fast Flow (high sub), is also available as lab packages and is an excellent choice for scale-up.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Débit:<4 ml/min
- Granulométrie:90 µm
- Capacité:1 ml
- Désignation:HiTrap™ Benzamidine FF (High Sub)
- Seq:01
- Matériau de colonne:PP
- Dimensions du lit:7×25 mm
- Plage de pH:2 - 8
- Cat. No.:17-5143-01
- Stockage:2 to 8 °C, 20% Ethanol, 0,05 M Sodium Acetate, pH 4,0
- Pression max.:5 bar



