Hydrometers, Brix
30 - 60 °Bx, 0,1 °Bx, 300 mm
N° de catalogue ALLA3910FC030/20QP
Fournisseur: Alla France
Aller à :
Product Details & Documents
These Brix sugar scale hydrometers are specially designed for use in breweries.
Spécifications
- Longueur:300 mm
- Division:0,1 °Bx
- Plage de mesure:30 - 60 °Bx