Hand-Drill, QualiRod
Échantillonneur manuel conique QualiRod, 175 mm, 120 mm
N° de catalogue 300-0018
Fournisseur: BURKLE
Aller à :
Product Details & Documents
Conical hand drill with sharp cutting edges and a solid handle made from stainless steel V2A (1.4301)/AISI 304.
- Sterilisable
Designed to cut out samples from soft, paste-like substances (cheese, butter, clay, loam, soap and wax).
: ISO 707
Spécifications
- Profondeur d'immersion:120 mm
- Désignation:Échantillonneur manuel conique QualiRod
- Longueur:175 mm



