Measuring cylinders, conical
Measuring cylinders, conical
500 ml, 115 mm, 115 mm
N° de catalogue BOCH8660
Fournisseur:   BOCHEM
Measuring cylinders, conical
N° de catalogue BOCH8660
Fournisseur:  BOCHEM
Measuring cylinders, conical
Measuring cylinders, conical
500 ml, 115 mm, 115 mm
Prix
24,80 €
Connexion Pour la disponibilité
Product Details & Documents
Stainless steel 18/10.
  • With base and handle
  • Conical, graduated
Spécifications
  • Ø:
    115 mm
  • Capacité:
    500 ml
  • Hauteur:
    115 mm