Visual immunoassays
Le produit 1.04140.0001 est similaire à 709-0253, mais le fabricant et les spécifications peuvent varier.
Product Details & Documents
Microtitre-format screening tests with high-affinity, specific antibodies for food and environment samples. Fast, specific bacteria and toxin testing with advanced Visual Immunoassay for food, water, environment samples, raw materials, cosmetics and pharmaceuticals. Ideal for rapid detection of pathogenic bacteria, 46 or 94 tests per plate.
- The simplicity of the VIA™ method frees up time for other work and projects
- Rapid results allow products to be released earlier, thereby saving on warehousing costs
- The kit can be used manually, semi automatically or fully automatically for large numbers of samples
- Fast results
- High sensitivity and specificity
Spécifications
Spécifications
- Cdt:96 Tests
- Désignation:Salmonella, VIA kit, 96 well