Adapters, receiver, plain bent, Quickfit®
24/29 NS, 24/29 NS
N° de catalogue 271-0016
Fournisseur:   SciLabware
Adapters, receiver, plain bent, Quickfit®
N° de catalogue 271-0016
Fournisseur:  SciLabware
Adapters, receiver, plain bent, Quickfit®
Adapters, receiver, plain bent, Quickfit®
24/29 NS, 24/29 NS
Prix
44,30 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Receiver, delivery, PYREX® borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Plain bend without vent.
Spécifications
  • Rodage mâle:
    24/29 NS
  • Rodage femelle:
    24/29 NS
  • Seq:
    04