Jars with base, knobbed lid
150 mm, 150 mm
N° de catalogue HECH44410036
Fournisseur: GLASWARENFABRIK KARL HECHT
Aller à :
Product Details & Documents
Made of glass, with glass or stainless steel lid.
- Good chemical resistance
- Sturdy
- Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Spécifications
- Ø ext.:150 mm
- Hauteur:150 mm