Jars with base, knobbed lid
120 mm, 120 mm
N° de catalogue 216-4295
Jars with base, knobbed lid
N° de catalogue 216-4295
Jars with base, knobbed lid
Jars with base, knobbed lid
120 mm, 120 mm
Prix
45,90 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Made of glass, with glass or stainless steel lid.
  • Good chemical resistance
  • Sturdy
  • Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Spécifications
  • Ø ext.:
    120 mm
  • Hauteur:
    120 mm