Jars with base, knobbed lid
Jars with base, knobbed lid
100 mm, 100 mm
Catalog # 216-4293
Jars with base, knobbed lid
Catalog # 216-4293
Jars with base, knobbed lid
Jars with base, knobbed lid
100 mm, 100 mm
Price
37,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of glass, with glass or stainless steel lid.
  • Good chemical resistance
  • Sturdy
  • Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Specifications
  • Ø ext.:
    100 mm
  • Hauteur:
    100 mm