Cover glasses
#1, 25 mm
N° de catalogue 631-9509
Fournisseur: GLASWARENFABRIK KARL HECHT
Aller à :
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
Informations de commande: Available in different diameters.
Spécifications
- Ø:25 mm
- Nb:#1
- Seq:13