Dissecting Forceps, Micro
Straight, with fine sharp tips, cross action, 100 mm
N° de catalogue HAMMHSO012-10
Fournisseur:   HAMMACHER KARL
Dissecting Forceps, Micro
N° de catalogue HAMMHSO012-10
Fournisseur:  HAMMACHER KARL
Dissecting Forceps, Micro
Dissecting Forceps, Micro
Straight, with fine sharp tips, cross action, 100 mm
Prix
103,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Extra fine micro forceps made from stainless steel.
Spécifications
  • Type:
    Straight, with fine sharp tips, cross action
  • Longueur:
    100 mm
  • Seq:
    03