Molybdenum standard solution
Molybdenum standard solution (NIST), 10 mg/l as Mo, 100 mL
  HACH1418742
 :   Hach
Molybdenum standard solution
  HACH1418742
 :  Hach
 :  
Molybdenum standard solution
Molybdenum standard solution
Molybdenum standard solution (NIST), 10 mg/l as Mo, 100 mL
 :  
43,40 €
1 * 100 mL
Standard solution for accuracy checks in molybdenum determinations.
  • Concentration:
    10 mg/l as Mo
  • Cdt:
    100 mL
  • Désignation:
    Molybdenum standard solution (NIST)