Molybdenum standard solution
Molybdenum standard solution (NIST), 10 mg/l as Mo, 100 mL
HACH1418742
: Hach
Standard solution for accuracy checks in molybdenum determinations.
- Concentration:10 mg/l as Mo
- Cdt:100 mL
- Désignation:Molybdenum standard solution (NIST)