Redox standards
Standard redox solution, 200 mV at 25 °C, 500 mL
Fournisseur: REAGECON
Aller à :
Product Details & Documents
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- Cdt:500 mL
- Désignation:Standard redox solution, 200 mV at 25 °C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



