D(+)-Mannose
1
Catalog # NENLNET-570A/001MC
Marketsource
Supplier: NEN LIFE SCIENCE
D(+)-Mannose
Catalog # NENLNET-570A/001MC
Supplier: NEN LIFE SCIENCE
Marketsource
CAS Number: 3458-28-4
Jump to:
Product Details & Documents
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Merck Index:13,05772
- Boiling Pt:376 °C (1013 hPa)
- Melting Pt:133 °C
- Density:1,539 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:222-392-4
Specifications
Specifications
- Cdt:1



