Eclipse™ Standard Surface Pipette Tips
Extended, Flextop, UltraFine, 1250 µl, +, 88,9 mm, Clear, Universal pipettes, 576/pack, 4608/case, 8 packs/case
N° de catalogue LBCN1045-965-008-9
New Product
Fournisseur: Labcon
Eclipse™ Standard Surface Pipette Tips
N° de catalogue LBCN1045-965-008-9
Fournisseur: Labcon
New Product
Aller à :
Product Details & Documents
Labcon's Eclipse™ standard surface pipette tips are made of high-quality, 100% pure virgin medical grade polypropylene. The resin is pre-tested to ensure no extractables are present.
Uniquely engineered to fit most popular brands of pipettes
- Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
- Tip sizes range from 10 to 1250 µl
- Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
- Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
- Made in USA
Labcon resins are pre-tested to ensure no extractables, PFAS, or other compounds are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs.
Certifications: PFAS-free, Endotoxin-free, ATP-free, RNase/Dnase-free, Low Trace Metals (<1 ppm).
Spécifications
- Emballage:576/pack, 4608/case, 8 packs/case
- Stérile:+
- Volume:1250 µl
- Longueur:88,9 mm
- Style de pointe:Extended, Flextop, UltraFine
- Couleur de pointe:Clear
- Pour l'utilisation avec:Universal pipettes