Pressure Bulbs
Ball set with net, merlot-red, size 5, natural rubber/PVC, 102
N° de catalogue DENE4120003
New Product
Fournisseur: DEUTSCH NEUMANN
Aller à :
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Spécifications
- Capacité:102
- Désignation:Ball set with net, merlot-red, size 5, natural rubber/PVC
- Seq:5