External Printer
Serial port RS232, for external printer
N° de catalogue SMEGWD-232PRINT
New Product
Fournisseur: SMEG
Aller à :
Product Details & Documents
Externla printer includes RS232 serial port card.
Dot matrix thermal printing
Complete with clock function
Equipped with independent power supply
For use with GW4260 series
To be combined with a roll of thermal paper Ø 50 mm and width 57 mm.
Spécifications
- Désignation:Serial port RS232, for external printer
- Seq:2