Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Saisie de commande
Login
/
Register
Produits
Services
Solutions numériques
Applications
Support
À propos
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
Lien pour commandes
ContactUsLinkComponent
Home
[EN]PAHS MIXTURE - 18 COMP.: 2000UG/ML E 1 * 1 mL
[EN]PAHS MIXTURE - 18 COMP.: 2000UG/ML E 1 * 1 mL
Print
[EN]PAHS MIXTURE - 18 COMP.: 2000UG/ML E 1 * 1 mL
Catalog # CMPLF223399
New Product
Supplier:
CUSTOM MADE CHEMICALS LAB
[EN]PAHS MIXTURE - 18 COMP.: 2000UG/ML E 1 * 1 mL
Catalog # CMPLF223399
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
Return to Top
[EN]PAHS MIXTURE - 18 COMP.: 2000UG/ML E 1 * 1 mL
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 mL
-
+
Inquire For Price
Return Policy
Product Details & Documents
Grade 595 ½ qualitative filter papers, Whatman™
#: 512-3622
Supplier: Whatman products (Cytiva)