External Printer
Imprimante externe
Catalog # SMEGWD-PRINTE2
New Product
Irregular Voltage
Supplier: SMEG
Jump to:
Product Details & Documents
Externla printer includes RS232 serial port card.
Dot matrix thermal printing
Complete with clock function
Equipped with independent power supply
For use with GW4260 series
To be combined with a roll of thermal paper Ø 50 mm and width 57 mm.
Specifications
- Désignation:Imprimante externe
- Cat. No.:SMEGWD-PRINTE2