Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Bromothymol Blue

Bottle
Catalog # APOSOR10125642.5KG
New Product
Supplier:   Apollo Scientific
Bromothymol Blue
Catalog # APOSOR10125642.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  
Bromothymol Blue

Bottle

CAS Number:  
Price
1 450,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • Molecular Weight:624,38 g/mol
  • EINECS:200-971-2
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications