Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

N-Benzylaniline

Bottle
Catalog # APOSOR350285-10KG
New Product
Supplier:   Apollo Scientific
N-Benzylaniline
Catalog # APOSOR350285-10KG
Supplier:  Apollo Scientific
New Product
CAS Number:  103-32-2
N-Benzylaniline

Bottle

CAS Number:  103-32-2
Price
3 650,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  N-Phenylbenzylamine, ((Phenylmethyl)amino)benzene
  • CAS Number:103-32-2
  • Formula:C₆H₅CH₂NHC₆H₅
  • MDL Number:MFCD00003018
  • Molecular Weight:183.25 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications