Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
4,4'-Dibromotriphenylamine

4,4'-Dibromotriphenylamine

Bottle
Catalog # APOSOR315393-25G
Irritant
New Product
Supplier:   Apollo Scientific
4,4'-Dibromotriphenylamine
Catalog # APOSOR315393-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  81090-53-1
4,4'-Dibromotriphenylamine

Bottle

CAS Number:  81090-53-1
Price
91,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N,N-Bis(p-bromophenyl)aniline
  • CAS Number:81090-53-1
  • Formula:C₁₈H₁₃Br₂N
  • MDL Number:MFCD00060104
  • Molecular Weight:403.12 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.