Sulphacetamide ≥98%
Bottle
N° de catalogue APOSBUP19458-5G
New Product
Fournisseur: Apollo Scientific
Sulphacetamide ≥98%
N° de catalogue APOSBUP19458-5G
Fournisseur: Apollo Scientific
New Product
Numéro CAS: 144-80-9
Aller à :
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymes: N-((4-Aminophenyl)sulphonyl)acetamide
- Numéro CAS:144-80-9
- Formule:C₈H₁₀N₂O₃S
- Poids moléculaire:214,24 g/mol
Spécifications
Spécifications
- Type de conditionnement:Bottle
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



