Mitomycine
Bottle
Catalog # APOSBIM0133-100MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Mitomycin C is a HIGHLY TOXIC antibiotic with antineoplastic properties.
Synonyms: Ametycin
- CAS Number:50-07-7
- Formula:C₁₅H₁₈N₄O₅
- Melting Pt:360 °C
- Storage Temperature:Réfrigérateur
- EINECS:200-008-6
- UN:3462
- ADR:6.1,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Type de conditionnement:Bottle