Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Adipohydrazide 97%

Adipohydrazide 97%

Bottle
Catalog # APOSOR1919-25G
New Product
Supplier:   Apollo Scientific
Adipohydrazide 97%
Catalog # APOSOR1919-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  1071-93-8
Adipohydrazide 97%

Bottle

CAS Number:  1071-93-8
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Adipyl hydrazide
  • CAS Number:1071-93-8
  • Formula:C₆H₁₄N₄O₂
  • MDL Number:MFCD00007614
  • Molecular Weight:174.2 g/mol
  • Melting Pt:180…182 °C
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications