EasyPlus™ Titrator Easy pH
Product Details & Documents
The Swiss engineered EasyPlus™ Titrator from METTLER TOLEDO offers a complete package for your routine application. Operation does not get simpler or more intuitive than this, thanks to the smartphone like iTitrate™ user interface and iTitrate™ intelligence.
- The Easy pH Titrator simplifies and speeds up automated acid/base endpoint or inflection/equivalence point titrations
Affordable Swiss technology; Swiss engineering and the highest quality parts and materials make the compact and accurate titrator a perfect addition to any quality laboratory. Quick start with iTitrate™ operation; Operation is simple with the Apps based user interface and intuitive system menu navigation; save training time and make daily tasks simpler and quicker.
Ready for your sample with iTitrate™ intelligence; The built-in intelligence requires only a few parameters to be set before the instrument is ready for titration. Support; Take advantage of our internet based support. Videos, FAQ’s and a multitude of applications are just a click away. Its choice of aqueous and non-aqueous pH electrodes makes this titrator ideal for the following applications and many more: Acidity content of juices, wines and milk; Alkalinity of cleaning reagents and chemicals; Free fatty acid content of cooking oils; Alkalinity (m-value) and acidity (p-value) of water.
Burette volume: 20 ml
Application repeatability: 0,5% RDS
Titration type: pH; Direct, back compensated, blank compensated
Livraison: Easy pH titrator include a sensor (EG11-BNC sensor pH aqueous), stirrer, 20 ml burette, tubing, operating instructions, installation and quick guide, declaration of conformity and test certificate.
Documents
EasyPlus - How to Perform a Titration EasyPlus Titrator - How to Determine Acidity Using an Autotitrator EasyPlus Titrator - How To Prepare Small Sample Volume EasyPlus Titrator - How To Prepare Solid Sample for Precipitation Titration
Vidéos
EasyPlus Easy pH, Easy Cl, Easy Ox, Easy Pro, Easy KFV Titrator (FR).pdf
Détails supplémentaires sur le produit
TDS EasyPlus Easy pH (FR).pdf
Fiche technique
Spécifications
- Désignation:Titrator EasyPlus™ Easy pH
- Includes:EG11-BNC sensor pH aqueous
- Cat. No.:METT30060041
- mV - Résolution:± 0,1 mV
- Connexions:1×USB-A, 1×USB-B; (EasyDirect PC connection with license)
- Gamme de température:0...100 °C
- Affichage:109,2 mm, 480×272 pixel RGB
- Résolution:± 0,1 °C
- Gamme de mesure:±2000 mV/0 - 14 pH
Accessories
Informations sur le produit
- N° de catalogueDésignationPlage de mesurePourDisponibilitéPrix
- METT30043104EG13-BNC sensor pH non-aqueous0 - 2000 mVEasy Pro, Easy KFV, Easy Ox titratorsConnexion Pour la disponibilité1 * 1 Pce - 336,00 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueCapacitéDésignationIntervalle de graduationDisponibilitéPrix
- 661-0175100 mlPlastic beaker (80 ml), 120 pcs.20 mlConnexion Pour la disponibilité1 * 120 ST - 113,00 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- METT30057635EasyPlus™ insert set titration head GTConnexion Pour la disponibilité1 * 1 Pce - 29,80 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- METT30065449EasyDirect instrument license for 3 titrators per PCConnexion Pour la disponibilité1 * 1 Pce - 956,00 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationPlage de mesurePourDisponibilitéPrix
- METT30043103EG11-BNC sensor pH aqueous±2000 mV/0 - 14 pHEasy Pro, Easy EPM, Easy pH titratorsConnexion Pour la disponibilité1 * 1 ST - 244,00 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix