DEPC ≥98%
New Product
Fournisseur: Apollo Scientific
Aller à :
À propos de cet article
Usage: Biochemical/life science applications
Synonymes: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- Numéro CAS:1609-47-8
- Formule:C₆H₁₀O₅
- Poids moléculaire:162,14 g/mol
2 Options disponibles ci-dessous
Product Details & Documents
Usage: Biochemical/life science applications
Synonymes: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- Numéro CAS:1609-47-8
- Formule:C₆H₁₀O₅
- Poids moléculaire:162,14 g/mol
Product Family Options
Informations sur le produit
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



