Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Emapunil ≥98%

Emapunil ≥98%

New Product
Supplier:   Apollo Scientific
Emapunil ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Emapunil, NVP-XBD173, XBD173, N-Ethyl-7,8-dihydro-7-methyl-8-oxo-2-phenyl-N-(phenylmethyl)-9H-purine-9-acetamide
  • CAS Number:226954-04-7
  • Formula:C₂₃H₂₃N₅O₂
  • Molecular Weight:401,46 g/mol
Product Family Options

Product Information

  • Type de conditionnement
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 351,00 €
    351,00 €
    1 * 25 mg
       351,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 694,00 €
    694,00 €
    1 * 50 mg
       694,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 999,00 €
    999,00 €
    1 * 100 mg
       999,00 €

    Specifications:
    New Product
    New Product