Iodine solution
Bottle
N° de catalogue APOSBIA182915-2.5L
Fournisseur: Apollo Scientific
Iodine solution
N° de catalogue APOSBIA182915-2.5L
Fournisseur: Apollo Scientific
Numéro CAS: 7553-56-2
Aller à :
Product Details & Documents
0,02365 mol/L (0,0473 N) (ASTM D 1510) volumetric solution.
Usage: Biochemical/life science applications
- Numéro CAS:7553-56-2
- Formule:I₂
- Poids moléculaire:253,81 g/mol
Spécifications
Spécifications
- Type de conditionnement:Bottle
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



