Stainless Steel Double-Ended Spatulas
Stainless Steel Double-Ended Spatulas
New Product
Fournisseur:   BURKLE
Stainless Steel Double-Ended Spatulas
Fournisseur:  BURKLE
New Product
Certificats
Stainless Steel Double-Ended Spatulas
9 Options disponibles ci-dessous
Product Details & Documents
Sturdy spatulas for precise dosing and gentle spreading.
  • Two edges: angular for precise work, rounded (curved) for gentle picking up and spreading
  • Made of high-quality stainless steel for durability, hygiene, and corrosion resistance
  • Versatile use; ideal for dosing, picking up, spreading, stirring, or mixing
This high-quality stainless steel double spatula is the ideal tool for precise and versatile everyday laboratory work. The spatula has a flat, angular edge on one side and a rounded (flat or curved) edge on the other. Whether portioning fine samples precisely, removing substances gently, or distributing larger quantities evenly, this spatula is suitable for every task. Thanks to its sophisticated geometry and solid stainless steel 1.4301 (304) construction, it is durable, hygienic and corrosion-resistant. An ideal tool for demanding applications in laboratories and industry.
The square edge is perfect for scraping, cutting or portioning solid or powdery substances, especially in the corners of cans or flat containers. The rounded side is ideal for removing, distributing, or stirring substances in round containers without scratching them. This double spatula replaces several individual tools, saving time and increasing work safety while reducing the risk of cross-contamination.
Emballage: Spatulas are packaged individually.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Longueur de lame
    Largeur de la lame
    Couleur
    Disponibilité
    Prix
  • Chattaway double-ended stainless steel spatula with square/round curved tip
    125 mm
    40 mm
    8 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 2,50 €
    2,50 €
    1 * 1 ST
       2,50 €
    Spécifications :
    New Product
    New Product
  • Chattaway double-ended stainless steel spatula with square/round curved tip
    150 mm
    45 mm
    8 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €
    Spécifications :
    New Product
    New Product
  • Chattaway double-ended stainless steel spatula with square/round curved tip
    184 mm
    50 mm
    10 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 3,00 €
    3,00 €
    1 * 1 ST
       3,00 €
    Spécifications :
    New Product
    New Product
  • Chattaway double-ended stainless steel spatula with square/round curved tip
    200 mm
    63 mm
    10 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 3,20 €
    3,20 €
    1 * 1 ST
       3,20 €
    Spécifications :
    New Product
    New Product
  • Chattaway double-ended stainless steel spatula with square/round curved tip
    237 mm
    66 mm
    11 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 3,70 €
    3,70 €
    1 * 1 ST
       3,70 €
    Spécifications :
    New Product
    New Product
  • Double-ended stainless steel spatula with angular/rounded tip
    150 mm
    45 mm
    22 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 2,30 €
    2,30 €
    1 * 1 ST
       2,30 €
    Spécifications :
    New Product
    New Product
  • Double-ended stainless steel spatula with angular/rounded tip
    210 mm
    60 mm
    22 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 3,80 €
    3,80 €
    1 * 1 ST
       3,80 €
    Spécifications :
    New Product
    New Product
  • Double-ended stainless steel spatula with angular/rounded tip
    300 mm
    75 mm
    22 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 13,40 €
    13,40 €
    1 * 1 ST
       13,40 €
    Spécifications :
    New Product
    New Product
  • Double-ended stainless steel spatula with angular/rounded tip
    500 mm
    100 mm
    22 mm
    argenté
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 21,60 €
    21,60 €
    1 * 1 ST
       21,60 €
    Spécifications :
    New Product
    New Product
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.