DIVTECH Fluogreen Lipid Nanoparticles for Cell Internalisation Control
DIVTECH Fluogreen Lipid Nanoparticles for Cell Internalisation Control
Fluogreen cell internalization nanoparticles (Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish), 1 ml
N° de catalogue DIVSDIV000F1
New Product
Fournisseur:   DIVERSA TECHNOLOGIES
DIVTECH Fluogreen Lipid Nanoparticles for Cell Internalisation Control
N° de catalogue DIVSDIV000F1
Fournisseur:  DIVERSA TECHNOLOGIES
New Product
Numéro CAS:  
DIVTECH Fluogreen Lipid Nanoparticles for Cell Internalisation Control
DIVTECH Fluogreen Lipid Nanoparticles for Cell Internalisation Control
Fluogreen cell internalization nanoparticles (Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish), 1 ml
Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
DIVERSA’s Cell internalisation uptake nanoparticles are a fluorescent, ready-to-use tool to monitor nanoparticle uptake and distribution in cells. They simplify the study of internalisation across cell types and experimental conditions, providing reproducible results in microscopy, flow cytometry, or plate-based assays. Biocompatible and stable, they help researchers validate delivery strategies before moving into therapeutic applications.
  • Fluorescently labeled nanoparticles for tracking
    Ideal for internalisation and biodistribution studies
    Suitable for 2D, 3D, and in vivo experiments, including immortalise cells, primary cells and organoids
    Stable in biological conditions
    Easy-to-use and ready-to-use formulation
    Non-toxic and biodegradable components
    Trackable by standard fluorescence detection platforms
Understanding how nanoparticles are internalised by cells is crucial for the design of effective delivery systems. The DIV000F1 kit provides fluorescently labeled, ready-to-use nanoparticles that mimic therapeutic formulations without carrying active cargo, making them ideal for uptake and biodistribution studies in 2D, 3D, and in vivo models.
Beyond direct visualisation, DIV000F1 serves as a standardised control to normalise cellular uptake capacity, allowing researchers to distinguish between biological differences and technical variability across cell lines or experimental conditions. This makes the kit particularly valuable in studies involving extracellular vesicles or exosomes, where it can act as a benchmark to validate internalisation results. Compatible with microscopy, flow cytometry, and plate-based assays, DIV000F1 provides a reliable, reproducible reference for optimizing nanoparticle delivery strategies.
Livraison: The kit contains one vial with the lipid mix for formulation. Each kit is provided with 1X DIVTECH empty vial for formulation preparation. Includes two pipette tips for 1 ml pipette to support accurate handling. Lot number and storage conditions indicated on every vial and box.
Attention: Products are intended for research use only (RUO).
Documents
Spécifications
Spécifications
  • Désignation:
    Fluogreen cell internalization nanoparticles (Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish)
  • Volume:
    1 ml
Spécifications
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.