External Printer
New Product
Supplier: SMEG
Jump to:
About this item
Externla printer includes RS232 serial port card.
Dot matrix thermal printing
Complete with clock function
Equipped with independent power supply
For use with GW4260 series
To be combined with a roll of thermal paper Ø 50 mm and width 57 mm.
2 Options Available Below
Product Details & Documents
Externla printer includes RS232 serial port card.
Dot matrix thermal printing
Complete with clock function
Equipped with independent power supply
For use with GW4260 series
To be combined with a roll of thermal paper Ø 50 mm and width 57 mm.
Product Family Options
Product Information
- DésignationAvailabilityPrice
- Specifications:Irregular VoltageNew Product
More
Specifications
Cat. No.SMEGWD-PRINTE2ModèleIrregular VoltageNew Product - Specifications:New Product
More
Specifications
Cat. No.SMEGWD-232PRINTModèleNew Product