Software, SW7000
Logiciel de communication entre l'équipement et le PC, SW7000, Affichage et enregistrement en temps réel, ou analyse post-cycle
N° de catalogue ESPISW7000
Fournisseur: RAYPA
Aller à :
Product Details & Documents
Allows the visualisation, documentation and printing of the processes carried out in the AE-DRY, AES, AHS-DRY and AHS-N series autoclaves.
Includes RS-232 cable, USB stick with software, and RS-232 to USB adapter
- Real-time and post-cycle data analysis
Communication software between the equipment and the PC for display and recording in real time or display after each cycle. Cycles can also be printed or exported to excel. PC connection via RS-232. Data can also be exported directly to a USB memory stick.
Spécifications
- Désignation:Logiciel de communication entre l'équipement et le PC, SW7000
- Pour:Affichage et enregistrement en temps réel, ou analyse post-cycle