Human estrogen (E) ELISA Kit
Human estrogen (E) ELISA Kit, 96 wells
N° de catalogue MSPPEK710943
Marketsource
Fournisseur: MARKET SOURCE PART PROCESS
Human estrogen (E) ELISA Kit
N° de catalogue MSPPEK710943
Fournisseur: MARKET SOURCE PART PROCESS
Marketsource
Numéro CAS:
Aller à :
Product Details & Documents
Spécifications
Spécifications
- Désignation:Human estrogen (E) ELISA Kit
- Tests par set:96 wells
- Type de test:Sandwich
- Méthode de détection:Colorimetric microplate reader
- Primary antibody reactivity:Human
- Portée de détection:3,3 - 200 pg/ml
- Type déchantillon:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Température de stockage:6 months (4 °C); 12 months (−20 °C)
- Cat. No.:MSPPEK710943
- Volume d'échantillon:50 - 100 µl
- Format:Pre-coated
- Cible:estrogen
- Sensibilité:0,5 pg/ml
- Regulatory status:RUO
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



