Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Nullscript ≥97% Inactive analog of scriptaid

Nullscript ≥97% Inactive analog of scriptaid

N° de catalogue ABCAAB141639-5
Marketsource
Fournisseur:   Abcam
Nullscript ≥97% Inactive analog of scriptaid
N° de catalogue ABCAAB141639-5
Fournisseur:  Abcam
Marketsource
Numéro CAS:  300816-11-9
Nullscript ≥97% Inactive analog of scriptaid
Numéro CAS:  300816-11-9
Prix
487,00 €
Connexion Pour la disponibilité
1 * 5 mg
Product Details & Documents

Inactive analog of histone deacetylase (HDAC) inhibitor scriptaid. Used as a negative control.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
  • Numéro CAS:300816-11-9
  • Poids moléculaire:298,3 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    100% Nullscript
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.