Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Dapsone ≥99% Antibacterial and anti-inflammatory agent

Dapsone ≥99% Antibacterial and anti-inflammatory agent

N° de catalogue ABCAAB141203-250
Fournisseur:   Abcam
Dapsone ≥99% Antibacterial and anti-inflammatory agent
N° de catalogue ABCAAB141203-250
Fournisseur:  Abcam
Numéro CAS:  80-08-0
Dapsone ≥99% Antibacterial and anti-inflammatory agent
Numéro CAS:  80-08-0
Prix
61,50 €
Connexion Pour la disponibilité
1 * 250 mg
Product Details & Documents

Antibacterial and anti-infla mMatory agent. Neuroprotective and anti-apoptotic effects.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • Numéro CAS:80-08-0
  • Poids moléculaire:248,3 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    100% Dapsone