Indigo carmine indicateur de pH, colorant pour histologie
À propos de cet article
pH indicator (blue at pH 11,4 and yellow at 13,0). Tissue stain. Also a tool for the determination of hypochlorite. Absorption: λmax 608 nm.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Numéro CAS:860-22-0
- Poids moléculaire:466,4 g/mol
2 Options disponibles ci-dessous
Product Details & Documents
pH indicator (blue at pH 11,4 and yellow at 13,0). Tissue stain. Also a tool for the determination of hypochlorite. Absorption: λmax 608 nm.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Numéro CAS:860-22-0
- Poids moléculaire:466,4 g/mol
Spécifications de la famille de produits
Résultats des tests de spécification
- Constituents::100% Indigo carmine



