Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
D-(+)-Cellobiose ≥99% beta-glucosidase substrate

D-(+)-Cellobiose ≥99% beta-glucosidase substrate

Marketsource
Fournisseur:   Abcam
D-(+)-Cellobiose ≥99% beta-glucosidase substrate
Fournisseur:  Abcam
Marketsource
Certificats

2 Options disponibles ci-dessous

Product Details & Documents

Disaccharide consisting of two β-glucose molecules linked by a β-1,4 glycosidic bond. Used as a substrate of β-glucosidase.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • Numéro CAS:528-50-7
  • Poids moléculaire:342,3 g/mol
Spécifications de la famille de produits

Résultats des tests de spécification

  • Constituents::
    100% D-(+) Cellobiose
Product Family Options

unology General

Informations sur le produit

  • Disponibilité
    Prix
  • Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 5 g
    1 * 5 g - 76,90 €
    76,90 €
    1 * 5 g
       76,90 €

    Spécifications :
    Marketsource
    Marketsource
  • Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 25 g
    1 * 25 g - 298,00 €
    298,00 €
    1 * 25 g
       298,00 €

    Spécifications :
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.