Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mitomycin solution 0,3% dans le DMSO ≥99% Anticancer and antibiotic agent

Mitomycin solution 0,3% dans le DMSO ≥99% Anticancer and antibiotic agent

N° de catalogue ABCAAB147469-2
Fournisseur:   Abcam
Mitomycin solution 0,3% dans le DMSO ≥99% Anticancer and antibiotic agent
N° de catalogue ABCAAB147469-2
Fournisseur:  Abcam
Numéro CAS:  50-07-7
Mitomycin solution 0,3% dans le DMSO ≥99% Anticancer and antibiotic agent
Numéro CAS:  50-07-7
Prix
66,70 €
Connexion Pour la disponibilité
1 * 2 mg
Product Details & Documents

Anticancer and antibiotic agent. Crosslinks DNA. Inhibits DNA synthesis and nuclear division. Also induces apoptosis.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  Ametycin
  • Numéro CAS:50-07-7
  • Poids moléculaire:334,33 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    99% Dimethylsulfoxide, 0,3% Mitomycin C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.