G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Fournisseur:   G-Biosciences
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Fournisseur:  G-Biosciences
Certificats
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
2 Options disponibles ci-dessous
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1.5 M sodium chloride
  • 0.5 M  sodium hydroxide
A high-pH reagent used to denature double-stranded nucleic acids into single strands, preparing DNA or RNA for hybridization. Ready-to-use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
Product Family Options
Informations sur le produit
  • Désignation
    Cdt
    Disponibilité
    Prix
  • Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH)
    1 L
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 L
    1 * 1 L - 79,00 €
    79,00 €
    1 * 1 L
       79,00 €
  • Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH)
    1 gallon
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 4 L
    1 * 4 L - 125,00 €
    125,00 €
    1 * 4 L
       125,00 €