Proteometer-CV Starter Kit Reagent (60)
Catalog # NOVIKIT-PCV-00101B
Supplier: NOVILYTIC
New Product
CAS Number:
Specifications
- Désignation:Proteometer-CV starter kit reagent (60)
Specifications
About this item
Fluorescent reagent portion of the Proteometer-CV starter kit. This is sold individually due to shipping and storage conditions.
- No sample preparation required - analysis directly from clarified fermentation broth without protein A purification
- Speed and efficiency - CV analysis in under 25 minutes
- Simplified workflow - all the components you need for seamless operation
- Exclusive technology - the only solution that performs charge variant analysis directly in clarified fermentation broth
- Store at −20 °C
Crimped, sealed, ready for use with the Proteometer-CV starter kit (NOVIKIT-PCV-00101A), which is purchased separately. This kit is reagent-only and pairs with our Proteometer-CV starter kit.
Reagent to be stored at −20 °C.
Ordering information: Please add starter Proteometer-CV kit seperately (NOVIKIT-PCV-001-01A).
Delivery:
Includes 1× Proteometer-CV fluorescent reagent vial. Allows for analysis up to 60 samples.
Packaging: Packaged separately from the Proteometer-CV full kit due to storage conditions.