KIMBLE® Polypropylene Racks for Vials
KIMBLE® Polypropylene Racks for Vials
Vial rack, depth 18 mm, 16 mm
N° de catalogue WHEA7492100016
Fournisseur:   DWK Life Sciences
KIMBLE® Polypropylene Racks for Vials
N° de catalogue WHEA7492100016
Fournisseur:  DWK Life Sciences
KIMBLE® Polypropylene Racks for Vials
Vial rack, depth 18 mm, 16 mm
Prix
478,00 €
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Ideal for the management of 12 mm and 16 mm OD vials within the laboratory.
  • Manufactured from polypropylene
    Lettered and numbered for indexing individual vials
    Interlocking corners for convenient stacking
Emballage: Case of ten.
Spécifications
  • Ø trou:
    16 mm
  • Désignation:
    Vial rack, depth 18 mm