Proteometer-UFT fluorescent reagent
Proteometer-UFT fluorescent reagent
Proteometer-UFT Fluorescent Reagent Vials (3X), to be stored at -20 °C
N° de catalogue NOVIRGN-PUF-001
New Product
Fournisseur:   NOVILYTIC
Proteometer-UFT fluorescent reagent
N° de catalogue NOVIRGN-PUF-001
Fournisseur:  NOVILYTIC
New Product
Numéro CAS:  
Proteometer-UFT fluorescent reagent
Proteometer-UFT fluorescent reagent
Proteometer-UFT Fluorescent Reagent Vials (3X), to be stored at -20 °C
Numéro CAS:  
Prix
1 150,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
3 vials of Proteometer-UFT Fluorescent reagent specifically formulated for use with Proteometer-UFT kit.
  • Vials of the Proteometer-UFT fluorescent reagent for reorder to continue analysis with the Proteometer-UFT kit
Intended for use with the Proteometer-UFT kit.
Informations de commande: Reorder in sets of 3 as needed.
Emballage: Supplied in a custom box with instructions for use.
Spécifications
Spécifications
  • Désignation:
    Proteometer-UFT Fluorescent Reagent Vials (3X), to be stored at -20 °C