Proteometer-L Consumables Kit (800 Titer)
Catalog # NOVIKIT-PL-005-01
Supplier: NOVILYTIC
New Product
CAS Number:
Specifications
- Désignation:Proteometer-L consumables kit
Specifications
About this item
800 Titer + aggregate assays. Consumables only, reactor not included. Designed for customers who are reordering. Use with any LC. For human mAbs, multi-specifics, recombinant proteins.
- mAb titer + aggregate measurement: one assay for two critical attributes
- No Protein A purification required: Reduces costs
- No sample pretreatment: Reduces hours of sample prep
- No method development: Reduces time until method useage
- Direct analysis from CFB: HCP removal not needed
- Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
- Low sample volume: reduces impact on processing
Eliminate sample prep and method development. The kit includes the Proteometer reagents, buffers, and other parts needed for a 'plug-and-play' use with any existing Proteometer-L including the starter kit. This kit does not include a Proteometer reactor.
Store at −20 °C.
Accessories information: See the Proteometer-L reactor, Proteometer-L buffer packets, and Proteometer-L reagent series for accessory component replacement options.
Ordering information: Glassware not included.
Delivery: Includes 10× Proteometer-L reagent vials, 10× Proteometer-L buffer packets, 1× reconstitution reagent - allows for analysis up to 800 samples.
Packaging: Supplied in a custom box with instructions.