Tork S5 Sensitive Hand Washing Mini Liquid Soap
Catalog # SCAA425702
Supplier: Essity
New Product
Specifications
- Emballage:8× 525 ml
- Désignation:Premium liquid soap, refill
Specifications
About this item
Tork sensitive hand washing mini liquid soap S5, allergy friendly, certified by ECARF, the European Centre for Allergy Research Foundation.
Recommended for more dirty hands that need more cleaning power, for example for use in kitchens
- Provides extra care for sensitive skin and nurtures dry skin with lipid-replenishing ingredients like glycerine, betaine and panthenol
- 96% of ingredients are from natural origin (according to specification and ISO 16128, including water).
-
Perfume free fragrance, premium quality
Colour: White
Compatible with S5 dispensers
Provide extra care for dry and sensitive skin with Tork sensitive liquid soap. With its skin-replenishing ingredients and no perfume, this soap cares for all skin types. This refill is suitable for Tork mini soap and sanitiser dispensers, easy to use, and provides good hand hygiene to all users.
Certifications:
EU Ecolabel, SE/030/002.