Spécifications
- Emballage:4 packs, 75 cloths each
- Couleur:blanc
- Désignation:Cloths, refill, premium, compatible with Tork W4 dispensers
- Longueur:108 mm (Folded size)
- L×l:415×355 mm (Un folded size)
Spécifications
À propos de cet article
Quickly deal with the most stubborn of spills, as this kitchen cloth is highly absorbent with a soft texture.
Avoid having to buy several types of cleaning cloth for kitchen tasks - this thick, cotton-based design can handle any mess
Safely and successfully clean hot surfaces with this heat-resistant cleaning cloth
Carry, open and dispose of packaging easily - these cleaning cloths come in a clever Tork Easy Handling® box
Keeping a busy kitchen clean is easy with these all-purpose kitchen cloths. Their thick, highly absorbent design can swiftly mop up tough spills. Plus, they're heat-resistant and stay strong even when wet for safe and reliable use. As these cleaning cloths are crafted from biobased fibres and come in packaging made from recycled materials, lower the environmental impact. When used with the single-sheet dispensing of Tork W4 cleaning cloth dispensers, they'll cut waste and promote hygiene, too. (TÜV Austria OK Biobased certified product)