ContactUsLinkComponent
b.safe Isolated Amplifier 1-Channel
b.safe Isolated Amplifier 1-Channel
Catalog # BOHLR660-04
Supplier:  Bohlender
New Product
undefined
b.safe Isolated Amplifier 1-Channel
Catalog # BOHLR660-04
Supplier:  Bohlender
New Product
Specifications
  • Désignation:
    1-channel isolated amplifier
  • l×p×h:
    200×200×40 mm
Specifications
About this item
Ready-to-install plastic housing made of ABS (IP66) protects against accidental contact with live contacts.
  • With clearly labelled housing screw connections to connect b.safe filling level sensors
  • 1-channel version for use with one approved level sensor
  • 2-channel for two approved level sensors and their safe operation in explosion-proof areas
Encapsulated housing for an installation, such as at the back wall of a safety cabinet. Safe signal exchange between explosion-hazardous and explosion-proof areas by galvanic isolation of the sensor side and the output side. In explosion-proof areas, the use of the b.safe Isolated Amplifier prevents possible ignitable sparks from the alarm electronics in the explosive atmosphere on the sensor side.
Combined with the b.safe Display or Mini-Display, the b.safe Isolated Amplifier enables an automatic recognition of sensors that are not connected or damaged, as well as line breaks. The use of the b.safe Isolated Amplifier with level controls, level sensors, and alarm boxes from other manufacturers must be checked on a case-by-case basis.
Ordering information:
Includes power supply (90-264 V/47-63 Hz) with plug adapter for EU, Australia, USA, Japan, and UK. Switch position factory set for use with b.safe System Waste Caps LC. The installation of b.safe Isolated Amplifier and the connection of the signal cable has to be carried out by qualified personnel.