Human Recombinant RAP1A (from E. coli)
À propos de cet article
Recombinant human RAP1A protein is a Human Full Length protein, in the 1 to 181 aa range, expressed in Escherichia coli, with ≥85% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 Options disponibles ci-dessous
Product Details & Documents
Recombinant human RAP1A protein is a Human Full Length protein, in the 1 to 181 aa range, expressed in Escherichia coli, with ≥85% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spécifications de la famille de produits
Spécifications
Spécifications
- Catalog No:
- ABCAAB97904-50
- ABCAAB97904-250
- Nom de la protéine/du peptide:
- RAP1A
- RAP1A
- Synonyme de protéine:
- KREV1|RAP1A|Ras-related protein Rap-1A|C21KG|G-22K|GTP-binding protein smg p21A|Ras-related protein Krev-1
- KREV1|RAP1A|Ras-related protein Rap-1A|C21KG|G-22K|GTP-binding protein smg p21A|Ras-related protein Krev-1
- Type de protéine/peptide:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugaison:
- Unconjugated
- Unconjugated
- Exempt de substance animale:
- N
- N
- Activité biologique:
- Standard
- Standard
- Purity:
- ≥85%
- ≥85%
- Concentration:
- 0,25 mg/ml
- 0,25 mg/ml
- Formulaire:
- Liquid
- Liquid
- Formulation:
- pH: 8, Constituents: 40% Glycerol (glycerin, glycerine), 1,16% Sodium chloride, 0,316% Tris HCl, 0,077% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- pH: 8, Constituents: 40% Glycerol (glycerin, glycerine), 1,16% Sodium chloride, 0,316% Tris HCl, 0,077% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Conditions de stockage:
- Store at −20 °C
- Store at −20 °C
- Température de transport:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Spécifications
Product Family Options
Informations sur le produit
- DisponibilitéPrix
- Spécifications :Marketsource
Plus
Specifications
Cat. No.ABCAAB97904-50Nom de la protéine/du peptideRAP1ASynonyme de protéineKREV1|RAP1A|Ras-related protein Rap-1A|C21KG|G-22K|GTP-binding protein smg p21A|Ras-related protein Krev-1Type de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedExempt de substance animaleNActivité biologiqueStandardPurity≥85%Concentration0,25 mg/mlFormulaireLiquidFormulationpH: 8, Constituents: 40% Glycerol (glycerin, glycerine), 1,16% Sodium chloride, 0,316% Tris HCl, 0,077% (R*,R*)-1,4-Dimercaptobutan-2,3-diolConditions de stockageStore at −20 °CTempérature de transport20Tested applicationsMS, SDS-PAGEMarketsource - Spécifications :Marketsource
Plus
Specifications
Cat. No.ABCAAB97904-250Nom de la protéine/du peptideRAP1ASynonyme de protéineKREV1|RAP1A|Ras-related protein Rap-1A|C21KG|G-22K|GTP-binding protein smg p21A|Ras-related protein Krev-1Type de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedExempt de substance animaleNActivité biologiqueStandardPurity≥85%Concentration0,25 mg/mlFormulaireLiquidFormulationpH: 8, Constituents: 40% Glycerol (glycerin, glycerine), 1,16% Sodium chloride, 0,316% Tris HCl, 0,077% (R*,R*)-1,4-Dimercaptobutan-2,3-diolConditions de stockageStore at −20 °CTempérature de transport20Tested applicationsMS, SDS-PAGEMarketsource



