Human Recombinant Rab3A (from E. coli)
À propos de cet article
Recombinant Human Rab3A protein is a Human Full Length protein, in the 1 to 220 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 Options disponibles ci-dessous
Product Details & Documents
Recombinant Human Rab3A protein is a Human Full Length protein, in the 1 to 220 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spécifications de la famille de produits
Spécifications
Spécifications
- Catalog No:
- ABCAAB109964-50
- ABCAAB109964-250
- Nom de la protéine/du peptide:
- Rab3A
- Rab3A
- Synonyme de protéine:
- Ras-related protein Rab-3A
- Ras-related protein Rab-3A
- Type de protéine/peptide:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugaison:
- Unconjugated
- Unconjugated
- Exempt de substance animale:
- N
- N
- Activité biologique:
- Standard
- Standard
- Purity:
- >90%
- >90%
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Formulaire:
- Liquid
- Liquid
- Formulation:
- pH: 8; Constituents: 20% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- pH: 8; Constituents: 20% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Conditions de stockage:
- Store at −20 °C
- Store at −20 °C
- Température de transport:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Spécifications
Product Family Options
Informations sur le produit
- DisponibilitéPrix
- Spécifications :
Plus
Specifications
Cat. No.ABCAAB109964-50Nom de la protéine/du peptideRab3ASynonyme de protéineRas-related protein Rab-3AType de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedExempt de substance animaleNActivité biologiqueStandardPurity>90%Concentration0,5 mg/mlFormulaireLiquidFormulationpH: 8; Constituents: 20% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolConditions de stockageStore at −20 °CTempérature de transport20Tested applicationsMS, SDS-PAGE - Spécifications :
Plus
Specifications
Cat. No.ABCAAB109964-250Nom de la protéine/du peptideRab3ASynonyme de protéineRas-related protein Rab-3AType de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedExempt de substance animaleNActivité biologiqueStandardPurity>90%Concentration0,5 mg/mlFormulaireLiquidFormulationpH: 8; Constituents: 20% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolConditions de stockageStore at −20 °CTempérature de transport20Tested applicationsMS, SDS-PAGE
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



