Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Human Recombinant Cripto1/CRIPTO (from HEK293 cells)

  ABCAAB84062-25
Marketsource
 :   Abcam
Human Recombinant Cripto1/CRIPTO (from HEK293 cells)
  ABCAAB84062-25
 :  Abcam
Marketsource
 :  
Human Recombinant Cripto1/CRIPTO (from HEK293 cells)
Human Recombinant Cripto1/CRIPTO (from HEK293 cells)
 :  
 
472,00 €
   
1 * 25 µG
 

Recombinant human Cripto1/CRIPTO protein (Fc Chimera) is a human fragment protein, in the 31 to 169 aa range, expressed in HEK 293, with >95% purity and suitable for FuncS, SDS-PAGE.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 

 

  • Cat. No.:
    ABCAAB84062-25
  • Conditions de stockage:
    –20 °C
  • Activité biologique:
    Standard
  • Exempt de substance animale:
    N
  • Purity:
    >95%
  • Synonyme de protéine:
    CRIPTO-1|TDGF1|CRIPTO|Protein Cripto|Cripto-1 growth factor|Epidermal growth factor-like cripto protein CR1|Teratocarcinoma-derived growth factor 1|CRGF
  • Conjugaison:
    Unconjugated
  • Tested applications:
    FuncS, SDS-PAGE
  • Species:
    Human
  • Nom de la protéine/du peptide:
    Cripto1/CRIPTO
  • Type de protéine/peptide:
    Recombinant
  • Source:
    HEK293 cells